Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RSK4 (aa 613-719) Control Fragment Recombinant Protein

Catalog No. RP92328
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP92328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP92328 Supplier Invitrogen™ Supplier No. RP92328
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82123. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

RPS6KA6 (RSK4) is a member of the RSK kinases family. RSK family members are activated by ERKs and mitogens and are unusual in that they contain 2 non-identical kinase domains. RSK4 encodes a member of ribosomal S6 kinase family, serine-threonine protein kinases which are regulated by growth factors. The encoded protein may be distinct from other members of this family, however, as studies suggest it is not growth factor dependent and may not participate in the same signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UK32
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 27330
Name Human RSK4 (aa 613-719) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2610524K04Rik; 90 kDa ribosomal protein S6 kinase 6; p90-RSK 6; p90RSK6; pp90RSK4; RGD1560817; ribosomal protein S6 kinase A6; ribosomal protein S6 kinase alpha-6; ribosomal protein S6 kinase polypeptide 6; ribosomal protein S6 kinase, 90kDa, polypeptide 6; ribosomal S6 kinase 4; Rps6ka6; RSK4; RSK-4; S6KA6; S6K-alpha 6; S6K-alpha-6
Common Name RSK4
Gene Symbol Rps6ka6
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YTMLAGYTPFANGPNDTPEEILLRIGNGKFSLSGGNWDNISDGAKDLLSHMLHMDPHQRYTAEQILKHSWITHRDQLPNDQPKRNDVSHVVKGAMVATYSALTHKTF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.