Learn More
Sino Biological Human RSPO1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 11083H08H1100
Encompass_Preferred
A DNA sequence encoding the amino acids (Met 1-Ala 146) of human RSPO1 (NP_001033722.1) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1188