Learn More
Sino Biological Human RSPO2 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 12507H02H100
Encompass_Preferred
A DNA sequence encoding the Human RSPO2 (Q6UXX9-1, with mutations L186P) (Met1-Gly205) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-4268