Learn More
Sino Biological Human RSV-F MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11049MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human respiratory syncytial virus (RSV) (A2) Fusion glycoprotein / RSV-F (Catalog# 11049-V08B; P03420.1; Met1-Thr529). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1238