Learn More
Sino Biological Human RYBP PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14292T26100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human RYBP (rh RYBP; Catalog#14292-H20B; Q8N488; Met1-Phe228). RYBP specific IgG was purified by Human RYBP affinity chromatography.
Catalog No. 50-237-0443