Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RyR3 (aa 1262-1338) Control Fragment Recombinant Protein

Catalog No. RP107118
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107118 Supplier Invitrogen™ Supplier No. RP107118
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84526. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

RYR3 is a ryanodine receptor, which functions to release calcium from intracellular storage for use in many cellular processes. RYR3 is involved in skeletal muscle contraction by releasing calcium from the sarcoplasmic reticulum followed by depolarization of T-tubules. Two transcript variants encoding different isoforms of RYR3 have been found. RYR3 mediates calcium channel release of Ca(2+) from the sarcoplasmic reticulum into the cytoplasm in muscle and plays a role in triggering muscle contraction. Further, RYR3 may also regulate Ca(2+) release by other calcium channels, as well as Ca(2+) release from the endoplasmic reticulum in non-muscle cells. RYR3 also contributes to cellular calcium ion homeostasis. Plays a role in cellular calcium signaling. Diseases associated with RYR3 include Central Core Disease and Arrhythmogenic Right Ventricular Dysplasia 2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15413
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 6263
Name Human RyR3 (aa 1262-1338) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI851294; Brain ryanodine receptor-calcium release channel; brain-type ryanodine receptor; C230090H21; calcium release channel; HBRR; Ryanodine receptor 3; RYR3; RYR-3; Type 3 ryanodine receptor
Common Name RyR3
Gene Symbol RYR3
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TMDSPPCLKVTHKTFGTQNSNADMIYCRLSMPVECHSSFSHSPCLDSEAFQKRKQMQEILSHTTTQCYYAIRIFAGQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.