Learn More
Raybiotech Inc Human S100 A6 ELISA Kit
Supplier:  Raybiotech Inc ELHS100A61
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.06 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human S100 A6.
Catalog No. 50-194-9694