Learn More
Biomatik Corporation Human S100 Calcium Binding Protein A5 (S100A5) ELISA Kit
Supplier:  Biomatik Corporation EKU07171
Encompass_Preferred
Human S100 Calcium Binding Protein A5 (S100A5) ELISA Kit for detecting S100A5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33763. Gene ID: 6276. Target Synonyms: S100-A5; S100D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7497