Learn More
Raybiotech Inc Human S100 P ELISA Kit
Supplier:  Raybiotech Inc ELHS100P1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 14 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human S100 P.
Catalog No. 50-194-9697