Learn More
Raybiotech Inc Human S100 A1 ELISA Kit
Supplier:  Raybiotech Inc ELHS100A11
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 0.82 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human S100A1. Other species not determined.
Catalog No. 50-194-9690