Learn More
Invitrogen™ Human S100A10 Recombinant Protein
Description
This product contains purified recombinant human S100A10 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P60903 |
| Concentration | 0.10 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | PBS with 50% glycerol and no preservative |
| Gene ID (Entrez) | 6281 |
| Molecular Weight (g/mol) | 15.3 kDa |
| Name | Human S100A10 |
| Quantity | 1 mg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.