Learn More
Sino Biological Anti-S100A11 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11140RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human S100A11 / S100C (rh S100A11; Catalog#11140-HNAE; NP_005611.1; Met1-Thr105). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4824