Learn More
Sino Biological Human S100A11 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11140T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human S100A11 / S100C (rh S100A11 / S100C; Catalog#11140-HNAE; NP_005611.1; Met1-Thr105). S100A11 / S100C specific IgG was purified by Human S100A11 / S100C affinity chromatography.
Catalog No. 50-237-1970