Learn More
Description
This product contains purified recombinant human S100A11 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
Specifications
Specifications
| Accession Number | P31949 |
| For Use With (Application) | Bioactivity, Western Blot |
| Formulation | 20mM tris HCl with 50% glycerol, 5mM EDTA and no preservative; pH 8 |
| Gene ID (Entrez) | 6282 |
| Name | Human S100A11 |
| pH Range | 8 |
| Purification Method | Purified |
| Quantity | 1 mg |
| Source | E. coli |
| Immunogen | N/A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.