Learn More
BIOHIPPO Human S100A12 Microsample
SDP
Catalog No. 5000313994
Encompass
Human S100A12 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of S100A12 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 30 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human S100A12 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human S100A12 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human S100A12 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13994 Supplier BIOHIPPO Supplier No. BHE15212329
Human S100A12 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of S100A12 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 30 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human S100A12 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human S100A12 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human S100A12 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color