Learn More
Sino Biological Human S100A13 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10991MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human S100A13 (rh S100A13; Catalog#10991-HNAE; NP_001019381.1; Met 1-Lys 98). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2822