Learn More
Sino Biological Anti-S100A6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10939RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human S100A6 (rh S100A6; Catalog#10939-H08B; NP_055439.1; Met1-Gly90). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-5045