Learn More
Sino Biological Anti-Psoriasin/S100A7 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11141MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human S100A7 (rh S100A7; Catalog#11141-HNAE; Met 1-Gln 101; NP_002954.2). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4823