Learn More
Sino Biological Recombinant Anti-Psoriasin/S100A7 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 11141R128F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A7 (rh S100A7; Catalog#11141-HNAE; NP_002954.2; Met1-Gln101) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-255-4819