Learn More
BIOHIPPO Human S100A7 Microsample
SDP
Catalog No. 5000315702
Encompass
Human S100A7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of S100A7 in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 53 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human S100A7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human S100A7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human S100A7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15702 Supplier BIOHIPPO Supplier No. BHE15211850
Human S100A7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of S100A7 in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 53 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human S100A7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human S100A7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human S100A7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color