Learn More
Sino Biological Anti-Psoriasin/S100A7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11141RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human S100A7 (rh S100A7; Catalog#11141-HNAE; NP_002954.2; Met 1-Gln 101). S100A7 specific IgG was purified by Human S100A7 affinity chromatography.
Catalog No. 50-255-4816