Learn More
Sino Biological Recombinant Anti-S100A8 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11138R203100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A8 (rh S100A8; Catalog#11138-H08B; NP_002955.2; Met1-Glu93).
Catalog No. 50-241-0339