Learn More
Sino Biological Anti-S100A8 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 11138MM07F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human S100A8 (rh S100A8; Catalog#11138-H08B; NP_002955.2; Met1-Glu93) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1303