Learn More
Sino Biological Human S100A8 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11138T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human S100A8 (rh S100A8; Catalog#11138-H08B; NP_002955.2; Met1-Glu93). S100A8 specific IgG was purified by Human S100A8 affinity chromatography.
Catalog No. 50-237-1418