Learn More
Sino Biological Recombinant Human S100A9 Protein (His Tag) 1
Supplier:  Sino Biological 11145H08B1
Encompass_Preferred
A DNA sequence encoding the human S100A9 (NP_002956.1) (Met 1-Pro 114) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7046