Learn More
Sino Biological Human S100A9 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11145R209100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A9 (rh S100A9; Catalog#11145-H08B; NP_002956.1; Met1-Pro114).
Catalog No. 50-237-1971