Learn More
Sino Biological Human S100B MAb 20ul (Sino Biological)
Supplier:  Sino Biological 68086MM2920
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human S100B (rh S100B; Catalog#10181-H07E; NP_006263.1; Ser2-Glu92). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-4098