Learn More
Sino Biological Recombinant Anti-S100B Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10181R227100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human S100B (rh S100B; Catalog#10181-H07E; NP_006263.1; Ser2-Glu92).
Catalog No. 50-240-8535