Learn More
Sino Biological Anti-S100B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100508T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human S100B, and purified by antigen affinity chromatography.
Catalog No. 50-240-9087