Learn More
Sino Biological Recombinant Human S100B Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10181H07E1
Encompass_Preferred
A DNA sequence encoding the human S100B (NP_006263.1) (Ser 2-Glu 92) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-240-9568