Learn More
Sino Biological Human SAA4 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 205686T08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SAA4 (rh SAA4; Catalog#11311-H09E; P35542; Glu19-Tyr130). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-4069