Learn More
Sino Biological Anti-SAA4 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11311MM03P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SAA4 (rh SAA4; Catalog#11311-H09E; P35542; Glu19-Tyr130) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8787