Learn More
Sino Biological Anti-SAA4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11311RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SAA4 (rh SAA4; Catalog#11311-H09E; P35542; Glu19-Tyr130). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8136