Learn More
Sino Biological Human SAP/SH2D1A Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785357
Encompass_Preferred
The Human SH2D1A ELISA Pair Set is for the quantitative determination of Human SH2D1A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human SH2D1A coated on a 96-well plate. Standards and samples are added to the wells, and any Human SH2D1A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human SH2D1A monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human SH2D1A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85357 Supplier Sino Biological Supplier No. SEK111495
The Human SH2D1A ELISA Pair Set is for the quantitative determination of Human SH2D1A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human SH2D1A coated on a 96-well plate. Standards and samples are added to the wells, and any Human SH2D1A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human SH2D1A monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human SH2D1A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.