Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SAPAP1 Synthetic Peptide

Catalog No. PEP0651 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0651 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0651 Supplier Invitrogen™ Supplier No. PEP0651
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 14 amino acids near the center of human SAPAP1. PEP-0651 can be used as a blocking peptide with polyclonal antibody PA5-20531.

Part of the postsynaptic scaffold in neuronal cells. Target binds the guanylate kinase-like domain of the PSD-95 family of synaptic ion channel clustering proteins; may play a role in protein-protein interactions at the synaptic junction.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 9229
Species Human
Purification Method purified
Accession Number O14490
Common Name SAPAP1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 4933422O14Rik; 9630002F18; AI845682; AI848168; BB075781; D17Bwg0511e; DAP1; DAP-1; DAP-1 beta; DAP-1-ALPHA; DAP-1-BETA; discs large homolog associated protein 1; discs, large (Drosophila) homolog-associated protein 1; disks large-associated protein 1; DLG associated protein 1; Dlgap1; DLGAP1A; DLGAP1B; GKAP; GKAP/SAPAP; GKAP/SAPAP1; GKPA/SAPAP; guanylate kinase associated protein; guanylate kinase-associated protein; hGKAP; Kiaa4162; mKIAA4162; PSD-95 binding protein; PSD-95/SAP90 binding protein 1; PSD-95/SAP90-binding protein 1; rGKAP; SAP90/PSD-95 associated protein 1; SAP90/PSD-95-associated protein 1; SAPAP1
Gene Symbol Dlgap1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.