Learn More
Biomatik Corporation Human Sarcoglycan Delta (SGCd) ELISA Kit
Supplier:  Biomatik Corporation EKU07193
Encompass_Preferred
Human Sarcoglycan Delta (SGCd) ELISA Kit for detecting SGCd in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.28ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92629. Gene ID: 6444. Target Synonyms: 35DAG; CMD1L; DAGD; SG-delta; SGCDP; SGD; 35kDa Dystrophin-Associated Glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7702