Learn More
STEMCELL Technologies Human SARS-CoV-2 Spike Protein Inhibitor Screening Kit, Size: 1 Kit
SDP
Supplier:  STEMCELL Technologies 1000700
Encompass
For detection and measurement of compounds that inhibit binding of viral SARS-CoV-2 Spike protein to human ACE2 receptor
Catalog No. 50-239-0171