Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SARS-CoV-2 Spike (Trimer) Ig Total ELISA Kit

Catalog No. BMS2323
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
2 x 96 Tests
5 x 96 Tests
96 Tests

ELISA

The Human SARS-CoV-2 Spike (Trimer) Ig Total ELISA is a serology assay that measures and quantitates immunoglobulin (Ig) antibodies against SARS-CoV-2 Spike (Trimer) in human serum or plasma. Principle of the method The Human SARS-CoV-2 Spike (Trimer) Ig Total ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of Ig antibodies bound to SARS-CoV-2 Spike (Trimer). A trimerized Spike protein is pre-coated in the wells of the supplied microplate. Samples, controls, including a high control that can be used as a standard are then added into these wells and bind to the immobilized (capture) Spike protein. The wells are washed and Ig antibodies conjugated to HRP are added and will bind to any captured antibodies. The Ig Total antibody recognizes IgG, IgM and IgA. The wells are washed and a substrate solution is added that reacts with the enzyme complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of antibody present in the original specimen.

It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV-2
Assay Range 62.5 to 4000 Units/mL
Assay Sensitivity <62.5 U/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 5%
Intraassay CV 4.3
Kit Contents Pre-coated 96 well plate, High Control (standard), Calibrator (medium control), Low Control, Anti-Ig Total - HRP conjugate, Assay Buffer concentrate, Wash Buffer concentrate, Substrate solution, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 5 μL, Serum 5 μL
Storage Requirements 2°C to 8°C
Target SARS-CoV-2 Spike (trimer) Protein
Test Time 30 mins
Total Assay Time 1 hrs 15 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.