Learn More
Biomatik Corporation Human SCAN Domain Containing Protein 3 (SCAND3) ELISA Kit
Supplier:  Biomatik Corporation EKU08379
Encompass_Preferred
Human SCAN Domain Containing Protein 3 (SCAND3) ELISA Kit for detecting SCAND3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6R2W3. Gene ID: 114821. Target Synonyms: ZNF452; ZFP38-L; ZBED9; Zinc finger BED domain-containing protein 9; Transposon-derived Buster4 transposase-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8672