Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SCAND1 (aa 145-179) Control Fragment Recombinant Protein

Catalog No. RP109255
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP109255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP109255 Supplier Invitrogen™ Supplier No. RP109255
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SCAND1 (SCAN domain containing 1), also known as RAZ1 or SDP1, is a nuclear protein that may play a role in the mechanism of transcription regulation. It is widely expressed with highest levels of expression found in kidney, thyroid, liver, prostate and testis. SCAND1 contains one SCAN box domain and, unlike the majority of other SCAN domain containing proteins, it does not contain a zinc finger motif. The SCAN box domain is a conserved leucine rich motif, approximately 60 amino acids in length, that participates in proteinprotein interactions. SCAND1 interacts with ZNF38, ZNF191 and MZF-1. It also interacts with ZNF202 and PPARgamma, suggesting that it plays a role in the transcriptional regulation of genes that are involved in energy homeostasis and lipid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P57086
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 51282
Name Human SCAND1 (aa 145-179) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2310003H23Rik; DIPP; DIPP1; Leap1; leucine-rich-domain inter-acting protein 1; peroxisome proliferative activated receptor, gamma, coactivator 2; PGC-2; Ppargc2; RAZ1; SCAN domain containing 1; SCAN domain-containing 1; SCAN domain-containing protein 1; Scand1; SCAN-related protein RAZ1; SDP1
Common Name SCAND1
Gene Symbol SCAND1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EQIVEMLVQEQLLAILPEAARARRIRRRTDVRITG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.