Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SCAPER (aa 85-180) Control Fragment Recombinant Protein

Catalog No. RP98089
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98089 Supplier Invitrogen™ Supplier No. RP98089
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61195. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CCNA2/CDK2 regulatory protein that transiently maintains CCNA2 in the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BY12
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 49855
Name Human SCAPER (aa 85-180) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias C430017I08; D530014O03Rik; KIAA1454; KIAA1454-like protein; LOW QUALITY PROTEIN: S phase cyclin A-associated protein in the endoplasmic reticulum; MSTP063; S phase cyclin A-associated protein in the endoplasmic reticulum; S phase cyclin A-associated protein in the ER; SCAPER; S-phase cyclin A associated protein in the ER; S-phase cyclin A-associated protein in the ER; Zfp291; Zinc finger protein 291; znf291
Common Name SCAPER
Gene Symbol SCAPER
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KSPTKTRHPRKIDLRARYWAFLFDNLRRAVDEIYVTCESDQSVVECKEVLMMLDNYVRDFKALIDWIQLQEKLEKTDAQSRPTSLAWEVKKMSPGR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.