Learn More
Sino Biological Anti-SR-BI/SCARB1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11069T38100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SR-BI/SCARB1/CD36L1 (rh SR-BI/SCARB1/CD36L1; Catalog#11069-H08H; NP_005496.4; Pro33-Tyr443). SR-BI/SCARB1/CD36L1 specific IgG was purified by Human SR-BI/SCARB1/CD36L1 affinity chromatography.
Catalog No. 50-255-4887