Learn More
ACROBiosystems ACROBiosystems Human SCARB1 / SR-B1 Protein, Fc Tag
Catalog No. 502106761
Encompass_Preferred
Recombinant Protein;1MG;Human SCARB1, Fc Tag (SC1-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 33 - Tyr 443 (Accession # NP_005496.4).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.7 kDa. As a result of glycosylation, the protein migrates as 110-115 kDa under reducing (R) condition, and 220-230 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-210-6761 Supplier ACROBiosystems Supplier No. SC1H52581MG
Recombinant Protein;1MG;Human SCARB1, Fc Tag (SC1-H5258) is expressed from human 293 cells (HEK293). It contains AA Pro 33 - Tyr 443 (Accession # NP_005496.4).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.7 kDa. As a result of glycosylation, the protein migrates as 110-115 kDa under reducing (R) condition, and 220-230 kDa under non-reducing (NR) condition (SDS-PAGE).