Learn More
Biomatik Corporation Human Scavenger Receptor Class D Member 1 (SCARD1) ELISA Kit
Supplier:  Biomatik Corporation EKU08435
Encompass_Preferred
Human Scavenger Receptor Class D Member 1 (SCARD1) ELISA Kit for detecting SCARD1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P34810. Gene ID: 968. Target Synonyms: CD68; GP110; Macrosialin; Macrophage Antigen CD68. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8666