Learn More
BIOHIPPO Human sCD130 Microsample
SDP
Catalog No. 5000313944
Encompass
Human sCD130 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sCD130 in Serum plasma and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 48 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human sCD130 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human sCD130 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human sCD130 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13944 Supplier BIOHIPPO Supplier No. BHE15210547
Human sCD130 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of sCD130 in Serum plasma and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 48 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human sCD130 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human sCD130 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human sCD130 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color