Learn More
Raybiotech Inc Human SCF R/CD117/c-Kit
Supplier:  Raybiotech Inc ELHSCFR1
Encompass_Preferred
Sensitivity: 45 pg/ml.Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human SCF R.
Catalog No. 50-194-9701