Learn More
Sino Biological Anti-Secretogranin II Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13441MM11100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Secretogranin II (rh Secretogranin II; Catalog#13441-H08H; AAH22509.1; Met1-Met617). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4239