Learn More
Sino Biological Anti-Secretogranin II Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13441RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SCG2 / Secretogranin II (rh SCG2 / Secretogranin II; Catalog#13441-H08H; AAH22509.1; Met1-Met617). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4238