Learn More
Sino Biological Human SCGN Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786626
Encompass_Preferred
The Human SCGN / Secretagogin ELISA Pair Set is for the quantitative determination of Human SCGN / Secretagogin.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for SCGN / Secretagogin coated on a 96-well plate. Standards and samples are added to the wells, and any SCGN / Secretagogin present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-SCGN / Secretagogin monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of SCGN / Secretagogin present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86626 Supplier Sino Biological Supplier No. SEK1241115
The Human SCGN / Secretagogin ELISA Pair Set is for the quantitative determination of Human SCGN / Secretagogin.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for SCGN / Secretagogin coated on a 96-well plate. Standards and samples are added to the wells, and any SCGN / Secretagogin present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-SCGN / Secretagogin monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of SCGN / Secretagogin present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.