Learn More
Biomatik Corporation Human Schlafen Family Member 5 (SLFN5) ELISA Kit
Supplier:  Biomatik Corporation EKU08405
Encompass_Preferred
Human Schlafen Family Member 5 (SLFN5) ELISA Kit for detecting SLFN5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q08AF3. Gene ID: 162394. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6028